Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodo-4-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A152210-5G A152210-100G A152210-10G A152210-25G A152210-5G
103842-528EA 6 USD InStock
103842-528 76768-178 76768-180 77318-144 77318-146
2-Iodo-4-(trifluoromethyl)aniline 98%
2-Iodo-4-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅F₃IN
MW: 287.02
Boiling Pt: 252.8 °C (760 mm Hg)
Melting Pt: 50 °C
Flash Pt: 106.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02683097
CAS Number: 163444-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty