Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-nitrophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A405914-1G A405914-5G A405914-1G A405914-5G A405914-100MG A405914-250MG
103827-128EA 30 USD InStock
103827-128 103838-168 77307-098 77307-100 77879-887 77879-888
4-Amino-3-nitrophenylboronic acid pinacol ester 98%
4-Amino-3-nitrophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₇BN₂O₄
MW: 264.09
Boiling Pt: 404.0±35.0 °C (760 mm Hg)
Melting Pt: 157.1…172.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06795680
CAS Number: 833486-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty