Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobutan-1-ol 97%

Supplier: AMBEED, INC
Danger

A916261-25G A916261-5G A916261-10G A916261-1G A916261-100G
103841-394EA 7 USD InStock
103841-394 76845-666 76845-668 77294-226 77308-518 77618-556
4-Aminobutan-1-ol 97%
4-Aminobutan-1-ol
Precaución: for Research Use Only


Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206
Melting Pt: 16…18 °C
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008230
CAS Number: 13325-10-5
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty