Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminoisoxazole hydrochloride 95%

Supplier: AMBEED, INC
Warning

A167491-1G A167491-5G A167491-100MG A167491-250MG A167491-1G
103806-192EA 165 USD InStock
103806-192 103844-454 76813-070 76813-072 77081-470
4-Aminoisoxazole hydrochloride 95%
4-Aminoisoxazole hydrochloride
Precaución: for Research Use Only


Formula: C₃H₅ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09992881
CAS Number: 108511-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty