Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminomethylquinoline hydrochloride 95%

Supplier: AMBEED, INC
Warning

A101950-1G A101950-5G A101950-100MG A101950-250MG
103796-218EA 195 USD InStock
103796-218 103821-912 77160-234 77160-236
4-Aminomethylquinoline hydrochloride 95%
4-Aminomethylquinoline hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClN₂
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD05861486
CAS Number: 1095661-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty