Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carboxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A109648-5G A109648-100G A109648-10G A109648-1G A109648-25G A109648-500G A109648-5G
103823-340EA 6 USD InStock
103823-340 77135-920 77135-922 77135-924 77135-926 77135-928 77310-150
4-Carboxyphenylboronic acid 98%
4-Carboxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BO₄
Boiling Pt: 406 °C (760 mm Hg)
Melting Pt: 220 °C
Flash Pt: 150.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00151801
CAS Number: 14047-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty