Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromo-1H-pyrazol-1-yl)ethanol 97%

Supplier: AMBEED, INC
Warning

A331394-1G A331394-5G A331394-250MG
103825-580EA 31 USD InStock
103825-580 103836-544 76839-534 77032-878
2-(4-Bromo-1H-pyrazol-1-yl)ethanol 97%
2-(4-Bromo-1H-pyrazol-1-yl)ethanol
Precaución: for Research Use Only


Formula: C₅H₇BrN₂O
MW: 191.03 g/mol
Boiling Pt: 299.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06804929
CAS Number: 214614-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty