Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1H-indole-7-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A115751-1G A115751-100MG A115751-250MG A115751-1G
103798-616EA 195 USD InStock
103798-616 103824-476 76982-950 76982-952 77311-294
4-Bromo-1H-indole-7-carboxylic acid 97%
4-Bromo-1H-indole-7-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₆BrNO₂
Boiling Pt: 455.8±30.0 °C (760 mm Hg)
Flash Pt: 229.5 °C
Storage Temperature: Ambient
MDL Number: MFCD12547329
CAS Number: 1211594-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty