Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A214908-100G A214908-10G A214908-500G A214908-1000G A214908-25G
103809-498EA 151 USD InStock
103809-498 103849-142 76999-738 76999-740 76999-742 77087-670 77087-672
4-Bromo-1H-pyrazole 98%
4-Bromo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₃BrN₂
Boiling Pt: 255 °C (760 mm Hg)
Melting Pt: 93…96 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00075602
CAS Number: 2075-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty