Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A119558-1G A119558-100MG A119558-5G A119558-25G
103799-782EA 23 USD InStock
103799-782 77026-296 77311-900 77871-516
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 97%
4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₆BrNO₂
MW: 204.0213 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07838483
CAS Number: 875160-43-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty