Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-naphthonitrile 98%

Supplier: AMBEED, INC
Warning

A663481-5G A663481-10G A663481-1G A663481-25G
103840-456EA 19 USD InStock
103840-456 76831-426 76844-648 76844-650
4-Bromo-1-naphthonitrile 98%
4-Bromo-1-naphthonitrile
Precaución: for Research Use Only


Formula: C₁₁H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD17012445
CAS Number: 92616-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty