Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,5-dichlorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A128739-1G A128739-250MG A128739-5G A128739-1G
103801-178EA 54 USD InStock
103801-178 76782-196 77298-322 77313-528
4-Bromo-2,5-dichlorobenzoic acid 97%
4-Bromo-2,5-dichlorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrCl₂O₂
MW: 269.91 g/mol
Boiling Pt: 347.574 °C (760 mm Hg)
Flash Pt: 164.007 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08444534
CAS Number: 885532-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty