Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-xylidine 98%

Supplier: AMBEED, INC
Warning

A561600-1000G A561600-100G A561600-10G A561600-500G A561600-25G
103810-452EA 187 USD InStock
103810-452 103839-878 76843-978 77197-636 77197-638 77197-640 77307-810
4-Bromo-2,6-xylidine 98%
4-Bromo-2,6-xylidine
Precaución: for Research Use Only


Formula: C₈H₁₀BrN
Boiling Pt: 258.8 °C (760 mm Hg)
Melting Pt: 48…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007826
CAS Number: 24596-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty