Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethyl-4-fluorobromobenzene 98%

Supplier: AMBEED, INC
Warning

A409793-1G A409793-5G A409793-100MG A409793-10G A409793-250MG A409793-1G A409793-25G A409793-5G
103827-200EA 13 USD InStock
103827-200 103838-248 76990-322 76990-324 76990-326 77307-136 77307-138 77307-140
3-Ethyl-4-fluorobromobenzene 98%
3-Ethyl-4-fluorobromobenzene
Precaución: for Research Use Only


Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD11846037
CAS Number: 627463-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty