Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-nitrophenol 95%

Supplier: AMBEED, INC
Danger

A112689-1G A112689-250MG A112689-1G
103798-076EA 59 USD InStock
103798-076 103823-916 76869-818 77297-200
4-Bromo-2-fluoro-5-nitrophenol 95%
4-Bromo-2-fluoro-5-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₃
Storage Temperature: Ambient
MDL Number: MFCD11100973
CAS Number: 661463-12-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty