Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-6-methylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A116068-1G A116068-100G A116068-10G A116068-250MG A116068-25G A116068-1G A116068-5G
103798-666EA 15 USD InStock
103798-666 103824-530 76810-182 76810-184 76810-186 76810-188 77311-354 77311-356
4-Bromo-2-fluoro-6-methylbenzoic acid 95%
4-Bromo-2-fluoro-6-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD18072908
CAS Number: 1242157-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty