Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorotoluene 98%

Supplier: AMBEED, INC
Warning

A158051-100G A158051-10G A158051-500G A158051-25G
103843-294EA 5 USD InStock
103843-294 76973-400 76973-402 76973-404 76973-406 77319-138
4-Bromo-2-fluorotoluene 98%
4-Bromo-2-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 68
Melting Pt: <20 °C
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty