Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-formylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A128036-1G A128036-100MG A128036-250MG
103801-056EA 47 USD InStock
103801-056 76822-818 76822-820
4-Bromo-2-formylbenzonitrile 97%
4-Bromo-2-formylbenzonitrile
Precaución: for Research Use Only


Formula: C₈H₄BrNO
MW: 210.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15144645
CAS Number: 713141-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty