Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-iodoanisole 98%

Supplier: AMBEED, INC
Warning

A124812-1G A124812-100G A124812-10G A124812-25G A124812-1G
103800-582EA 17 USD InStock
103800-582 76983-424 76983-426 76983-428 77312-724
4-Bromo-2-iodoanisole 98%
4-Bromo-2-iodoanisole
Precaución: for Research Use Only


Formula: C₇H₆BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD13186707
CAS Number: 98273-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty