Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-nitroanisole 98%

Supplier: AMBEED, INC
Warning

A113901-1G A113901-5G A113901-100G A113901-10G A113901-250MG A113901-25G A113901-1G A113901-5G
103798-316EA 11 USD InStock
103798-316 103824-158 76995-382 76995-384 76995-386 76995-388 77310-972 77310-974
5-Bromo-2-nitroanisole 98%
5-Bromo-2-nitroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrNO₃
MW: 232.03
Boiling Pt: 298.7±20.0 °C (760 mm Hg)
Melting Pt: 90…95 °C
Flash Pt: 134.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09966092
CAS Number: 103966-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty