Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-methoxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A271731-1G A271731-5G A271731-25G A271731-5G A271731-100G A271731-10G A271731-1G
103813-060EA 9 USD InStock
103813-060 103852-972 77092-754 77092-756 77289-458 77289-460 77305-094
4-Bromo-2-methoxybenzoic acid 98%
4-Bromo-2-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MW: 231.05
Boiling Pt: 318 °C (760 mm Hg)
Melting Pt: 155…159 °C
Flash Pt: 146.168 °C
Storage Temperature: Ambient
MDL Number: MFCD07780665
CAS Number: 72135-36-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty