Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-methoxybenzonitrile 97%

Supplier: AMBEED, INC
Warning

A208078-1G A208078-25G A208078-5G A208078-100G A208078-10G A208078-1G
103817-852EA 5 USD InStock
103817-852 103848-538 77086-840 77086-842 77164-874 77164-876 77303-056
4-Bromo-2-methoxybenzonitrile 97%
4-Bromo-2-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆BrNO
Boiling Pt: 289.1 °C (760 mm Hg)
Flash Pt: 128.66 °C
Storage Temperature: Ambient
MDL Number: MFCD06738651
CAS Number: 330793-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty