Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A177877-5G A177877-100G A177877-1G
103845-614EA 13 USD InStock
103845-614 76998-458 76998-460
4-Bromo-2-methylpyridine 98%
4-Bromo-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₆BrN
MW: 172.02
Boiling Pt: 195.7 °C (760 mm Hg)
Melting Pt: 26…27 °C (760 mm Hg)
Flash Pt: 72.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788196
CAS Number: 22282-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty