Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-dimethyl-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A264548-5G A264548-100G A264548-10G A264548-500G A264548-25G A264548-5G
103852-578EA 5 USD InStock
103852-578 77014-248 77014-250 77014-252 77092-244 77092-246
4-Bromo-3,5-dimethyl-1H-pyrazole 95%
4-Bromo-3,5-dimethyl-1H-pyrazole


Formula: C₅H₇BrN₂
Boiling Pt: 341 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00005242
CAS Number: 3398-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty