Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A116372-1G A116372-100G A116372-10G A116372-25G A116372-1G A116372-5G
103798-724EA 9 USD InStock
103798-724 103824-584 76766-208 76766-210 76766-212 77297-456 77297-458
4-Bromo-3-fluorobenzaldehyde 98%
4-Bromo-3-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄BrFO
Melting Pt: 55…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095000
CAS Number: 133059-43-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty