Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorobenzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A236535-1G A236535-5G A236535-25G A236535-5G A236535-10G A236535-1G
103811-106EA 6 USD InStock
103811-106 103850-878 77089-928 77089-930 77153-012 77304-054
4-Bromo-3-fluorobenzenesulphonyl chloride 98%
4-Bromo-3-fluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃BrClFO₂S
Boiling Pt: 277.3 °C (760 mm Hg)
Flash Pt: 121.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03094150
CAS Number: 351003-51-5
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty