Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-fluoro-4-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A327671-25G A327671-5G A327671-100G A327671-10G
103836-420EA 11 USD InStock
103836-420 76839-388 76839-390 77290-374 77290-376
1-Bromo-2-fluoro-4-nitrobenzene 97%
1-Bromo-2-fluoro-4-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 252.9 °C (760 mm Hg)
Melting Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD05865088
CAS Number: 185331-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty