Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-isopropylbenzonitrile 95%

Supplier: AMBEED, INC
Warning

A1001421-1G A1001421-5G A1001421-100MG A1001421-1G A1001421-5G
103795-268EA 177 USD InStock
103795-268 103820-986 76645-882 77282-440 77282-442
4-Bromo-3-isopropylbenzonitrile 95%
4-Bromo-3-isopropylbenzonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrN
MW: 224.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094529
CAS Number: 1369954-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty