Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-nitrobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A103310-1G A103310-5G A103310-100G A103310-25G
103796-526EA 5 USD InStock
103796-526 103822-230 76765-310 76765-312
4-Bromo-3-nitrobenzoic acid 97%
4-Bromo-3-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄BrNO₄
Boiling Pt: 340.9 °C (760 mm Hg)
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00272137
CAS Number: 6319-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty