Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-fluoro-2-nitrobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A474733-5G A474733-1G A474733-25G A474733-100G A474733-10G A474733-250MG
103828-066EA 6 USD InStock
103828-066 103839-138 76843-066 76843-068 77116-386 77116-388 77116-390
4-Bromo-5-fluoro-2-nitrobenzoic acid 98%
4-Bromo-5-fluoro-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrFNO₄
Boiling Pt: 367.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08702773
CAS Number: 1020717-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty