Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-methylpicolinic acid 95%

Supplier: AMBEED, INC
Warning

A112517-1G A112517-100MG A112517-250MG A112517-1G A112517-5G
103798-028EA 159 USD InStock
103798-028 103823-864 76821-936 76821-938 77297-186 77297-188
4-Bromo-5-methylpicolinic acid 95%
4-Bromo-5-methylpicolinic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13190305
CAS Number: 1196154-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty