Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-methoxy-1H-indole 98%

Supplier: AMBEED, INC
Warning

A171194-1G A171194-5G A171194-100MG A171194-10G A171194-250MG A171194-1G A171194-5G
103806-602EA 183 USD InStock
103806-602 103844-876 77286-858 77286-860 77286-862 77301-102 77301-104
4-Bromo-6-methoxy-1H-indole 98%
4-Bromo-6-methoxy-1H-indole
Precaución: for Research Use Only


Formula: C₉H₈BrNO
MW: 226.07 g/mol
Boiling Pt: 346.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03095207
CAS Number: 393553-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty