Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-7-fluoroindan-1-one 97%

Supplier: AMBEED, INC
Warning

A172099-1G A172099-5G A172099-100G A172099-100MG A172099-250MG A172099-25G A172099-1G A172099-5G
103806-696EA 14 USD InStock
103806-696 103844-980 76824-538 76824-540 76824-542 76824-544 77082-158 77082-160
4-Bromo-7-fluoroindan-1-one 97%
4-Bromo-7-fluoroindan-1-one
Precaución: for Research Use Only


Formula: C₉H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD17677340
CAS Number: 1003048-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty