Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-7-methoxy-1H-indole 95%

Supplier: AMBEED, INC
Warning

A550547-1G A550547-250MG A550547-5G A550547-100MG
103828-690EA 99 USD InStock
103828-690 103839-818 76819-052 76843-888 77882-330
4-Bromo-7-methoxy-1H-indole 95%
4-Bromo-7-methoxy-1H-indole
Precaución: for Research Use Only


Formula: C₉H₈BrNO
MW: 226.0700 g/mol
Boiling Pt: 356.49 °C (760 mm Hg)
Flash Pt: 169.4 °C
Storage Temperature: Ambient
MDL Number: MFCD08272257
CAS Number: 436091-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty