Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzenesulfonamide 97%

Supplier: AMBEED, INC
Warning

A139876-10G A139876-1G A139876-5G A139876-25G
77150-014EA 11 USD InStock
77150-014 77150-016 77299-092 77315-890
4-Bromobenzenesulfonamide 97%
4-Bromobenzenesulphonamide
Precaución: For research use only.


Formula: C₆H₆BrNO₂S
Boiling Pt: 360.7 °C (760 mm Hg)
Melting Pt: 163…167 °C
Flash Pt: 171.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00051977
CAS Number: 701-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty