Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromooxindole 97%

Supplier: AMBEED, INC
Warning

A389690-5G A389690-1G A389690-25G A389690-5G A389690-100G A389690-100MG A389690-10G A389690-250MG
103826-850EA 9 USD InStock
103826-850 103837-890 76841-254 76841-256 76841-258 77155-432 77155-434 77155-436 77155-438
4-Bromooxindole 97%
4-Bromooxindole
Precaución: for Research Use Only


Formula: C₈H₆BrNO
Boiling Pt: 358.2 °C (760 mm Hg)
Melting Pt: 202…208 °C
Flash Pt: 170.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06659911
CAS Number: 99365-48-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty