Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-formylpyridine 97%

Supplier: AMBEED, INC
Warning

A100505-1G A100505-5G A100505-10G A100505-250MG A100505-25G A100505-1G A100505-5G
103795-486EA 21 USD InStock
103795-486 103821-180 76780-374 76780-376 76780-378 77296-730 77296-732
4-Bromo-2-formylpyridine 97%
4-Bromo-2-formylpyridine
Precaución: for Research Use Only


Formula: C₆H₄BrNO
Boiling Pt: 243.9 °C (760 mm Hg)
Flash Pt: 101.354 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690697
CAS Number: 131747-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty