Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromothiophenol 98%

Supplier: AMBEED, INC
Danger

A711689-5G A711689-25G A711689-5G A711689-10G
103840-702EA 5 USD InStock
103840-702 76844-918 76844-920 77293-334
4-Bromothiophenol 98%
4-Bromothiophenol
Precaución: for Research Use Only


Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 239
Melting Pt: 71…76 °C
Flash Pt: 93.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004845
CAS Number: 106-53-6
UN: 3335
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty