Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A236883-100G A236883-500G A236883-1000G A236883-5G A236883-25G
103809-672EA 220 USD InStock
103809-672 103850-894 76771-294 76771-296 77089-956 77089-958 77304-066
4-Butylbenzoic acid 98%
4-Butylbenzoic acid
Precaución: For research use only.


Formula: C₁₁H₁₄O₂
Boiling Pt: 300.2 °C (760 mm Hg)
Melting Pt: 100…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002571
CAS Number: 20651-71-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty