Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amidinobenzoic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A105611-1G A105611-5G A105611-250MG A105611-1G
103796-908EA 37 USD InStock
103796-908 103822-642 77093-738 77309-510
4-Amidinobenzoic acid hydrochloride 95%
4-Amidinobenzoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₈N₂O₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD00269643
CAS Number: 42823-72-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty