Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroimidazo[4,5-c]pyridine 97%

Supplier: AMBEED, INC
Warning

A232208-1G A232208-5G A232208-1G A232208-25G A232208-5G A232208-10G A232208-250MG
103810-822EA 21 USD InStock
103810-822 103850-574 77089-498 77089-500 77089-502 77288-434 77288-436
4-Chloroimidazo[4,5-c]pyridine 97%
4-Chloroimidazo[4,5-c]pyridine
Precaución: for Research Use Only


Formula: C₆H₄ClN₃
Boiling Pt: 455.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08689582
CAS Number: 2770-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty