Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A583201-1G A583201-5G A583201-1G A583201-5G
103828-872EA 22 USD InStock
103828-872 103840-022 77307-870 77307-872
4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde 95%
4-Chloro-2-(methylthio)pyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₆H₅ClN₂OS
MW: 188.635 g/mol
Boiling Pt: 315 °C (760 mm Hg)
Flash Pt: 144 °C
Storage Temperature: Freezer
MDL Number: MFCD16657918
CAS Number: 148256-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty