Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(methylsulfonyl)pyrimidine 98%

Supplier: AMBEED, INC
Warning

A144270-1G A144270-5G A144270-250MG A144270-1G A144270-5G
103803-384EA 12 USD InStock
103803-384 103841-460 77285-866 77316-700 77316-702
4-Chloro-2-(methylsulfonyl)pyrimidine 98%
4-Chloro-2-(methylsulfonyl)pyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O₂S
Boiling Pt: 371.1 °C (760 mm Hg)
Melting Pt: 87…89 °C
Flash Pt: 178.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09701372
CAS Number: 97229-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty