Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A331627-5G A331627-1G A331627-25G A331627-10G A331627-250MG A331627-100G
103825-582EA 6 USD InStock
103825-582 103836-546 76839-536 76839-538 77099-992 77099-994 77685-949
4-Chloro-2-fluorophenylboronic acid 97%
4-Chloro-2-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BClFO₂
Boiling Pt: 289.2 °C (760 mm Hg)
Melting Pt: 248…250 °C
Flash Pt: 128.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02684293
CAS Number: 160591-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty