Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A179126-1G A179126-5G A179126-1G A179126-5G A179126-250MG
103807-398EA 6 USD InStock
103807-398 103845-740 77083-188 77083-190 77163-810
4-Chloro-2-hydroxypyridine 98%
4-Chloro-2-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₄ClNO
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 177…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093022
CAS Number: 40673-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty