Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-methoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A198268-1G A198268-5G A198268-1G A198268-5G A198268-10G A198268-250MG
103817-008EA 32 USD InStock
103817-008 103847-634 77085-654 77085-656 77287-616 77287-618
4-Chloro-2-methoxybenzaldehyde 97%
4-Chloro-2-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Melting Pt: 70…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703355
CAS Number: 53581-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty