Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A128410-1G A128410-10G A128410-25G A128410-5G A128410-1G
103801-120EA 52 USD InStock
103801-120 76822-836 77298-300 77298-302 77313-422
4-Chloro-3-hydroxypyridine 98%
4-Chloro-3-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₄ClNO
Boiling Pt: 298.2 °C (760 mm Hg)
Flash Pt: 134.1 °C
Storage Temperature: Ambient
MDL Number: MFCD02682915
CAS Number: 96630-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty