Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-methoxybenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A245677-1G A245677-5G A245677-100G A245677-25G A245677-5G A245677-1G
103811-662EA 7 USD InStock
103811-662 103851-494 76815-512 77090-758 77090-760 77304-364
4-Chloro-3-methoxybenzaldehyde 98%
4-Chloro-3-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
Boiling Pt: 265 °C (760 mm Hg)
Melting Pt: 56…60 °C
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787490
CAS Number: 13726-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty