Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-methylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A120597-1G A120597-10G A120597-250MG A120597-1G
103799-966EA 37 USD InStock
103799-966 77108-558 77108-560 77312-072
4-Chloro-5-methylpyrimidine 97%
4-Chloro-5-methylpyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂
Boiling Pt: 195.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08236809
CAS Number: 51957-32-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty