Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-(trifluoromethyl)quinazoline 95%

Supplier: AMBEED, INC
Warning

A494799-100MG A494799-1G A494799-250MG
103828-244EA 108 USD InStock
103828-244 103839-322 76789-850 76843-288 77669-220
4-Chloro-6-(trifluoromethyl)quinazoline 95%
4-Chloro-6-(trifluoromethyl)quinazoline
Precaución: for Research Use Only


Formula: C₉H₄ClF₃N₂
Boiling Pt: 285.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09837194
CAS Number: 16499-64-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty